Learn More
Biomatik Corporation Csga Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11911
Encompass_Preferred
Csga Polyclonal Antibody is a Escherichia coli O157:H7 Polyclonal Antibody to CSGA. The Csga Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli O157:H7. Immunogen: Recombinant Escherichia Coli O157:H7 Csga Protein. Applications: ELISA, WB. Target Synonyms: CSGA. Accession Number: Q93U24. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5688