Learn More
Waters Corp Method Transfer Kit, CSH C18, 130Å, 1.7–5 μm, 2 pk
Catalog No. 50791486 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Transfer Kit includes CSH and matching HPLC columns, engineered for identical selectivity and length-to-particle size ratios. They offer alternative reversed-phase selectivity, improving peak shape and loading capacity for basic compounds, minimizing column bleed, and ensuring rapid mobile-phase re-equilibration. Charged Surface Hybrid (CSH) technology provides exceptional peak shape in low-ionic-strength mobile phases and supports MS-friendly method development without using TFA.
  • Optimized peak shape and loading capacity for basic compounds.
  • Rapid column equilibration and consistent results.
  • MS-friendly method development (TFA-like performance without TFA).
  • Enhanced resolution, sensitivity, and analysis speed.
  • High pressure tolerance (up to 18000 psi) and extended pH stability.
  • Seamless UPLC-HPLC transfer and excellent reproducibility.
Catalog No. 50-791-486 Supplier Waters Corp Supplier No. 186005529
This Method Transfer Kit includes CSH and matching HPLC columns, engineered for identical selectivity and length-to-particle size ratios. They offer alternative reversed-phase selectivity, improving peak shape and loading capacity for basic compounds, minimizing column bleed, and ensuring rapid mobile-phase re-equilibration. Charged Surface Hybrid (CSH) technology provides exceptional peak shape in low-ionic-strength mobile phases and supports MS-friendly method development without using TFA.
  • Optimized peak shape and loading capacity for basic compounds.
  • Rapid column equilibration and consistent results.
  • MS-friendly method development (TFA-like performance without TFA).
  • Enhanced resolution, sensitivity, and analysis speed.
  • High pressure tolerance (up to 18000 psi) and extended pH stability.
  • Seamless UPLC-HPLC transfer and excellent reproducibility.