Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Column, 130Å, 1.7 μm
Catalog No. NC9052927 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These CSH C18 columns provide alternative selectivity and ultra-efficient separations. They address common issues like peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides alternate selectivity and ultra-efficiency for reversed-phase UPLC.
  • Addresses peak shape asymmetry and enhances loading capacity for basic compounds.
  • Minimizes column bleed and ensures rapid mobile-phase re-equilibration.
  • Separates a broad spectrum of compounds due to robust design and wide pH stability.
  • Provides high-resolution, high-efficiency chromatography for fast UPLC analysis.
  • Offers flexibility in method development and compatibility with biological samples.
Catalog No. NC9052927 Supplier Waters Corp Supplier No. 186005298
These CSH C18 columns provide alternative selectivity and ultra-efficient separations. They address common issues like peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and slow mobile-phase re-equilibration.
  • Provides alternate selectivity and ultra-efficiency for reversed-phase UPLC.
  • Addresses peak shape asymmetry and enhances loading capacity for basic compounds.
  • Minimizes column bleed and ensures rapid mobile-phase re-equilibration.
  • Separates a broad spectrum of compounds due to robust design and wide pH stability.
  • Provides high-resolution, high-efficiency chromatography for fast UPLC analysis.
  • Offers flexibility in method development and compatibility with biological samples.