Learn More
Aviva Systems Biology Corp CSH1 Antibody - middle region (ARP40350_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40350_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CSH1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CSH1. Alternate names: PL, CSA, CS-1, CSMT, hCS-AApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53192