Learn More
Enzo Life Sciences CSH103 substrate (500µg). CAS: 135546-44-0
SDP
Catalog No. 502011226
Encompass_Preferred
This sequence, ADAQHATPPKKKRKVED, is SV40 L Tag (118-138) with Ser to Ala120 and Ala123 substitutions to increase specificity. Substrate is specific for p34cdc2 (CDK1) and is a poor substrate for CKI, II, PKA, GSK3, PKC, CaMKII, and phosphorylase b kinase. Can be used in cell lysates using TCA/phosphocellulose assay. Km=74 µM for active complex with cyclin B. Long Term Storage: -20°C.
Catalog No. 50-201-1226 Supplier Enzo Life Sciences Supplier No. BMLP1040500
This sequence, ADAQHATPPKKKRKVED, is SV40 L Tag (118-138) with Ser to Ala120 and Ala123 substitutions to increase specificity. Substrate is specific for p34cdc2 (CDK1) and is a poor substrate for CKI, II, PKA, GSK3, PKC, CaMKII, and phosphorylase b kinase. Can be used in cell lysates using TCA/phosphocellulose assay. Km=74 µM for active complex with cyclin B. Long Term Storage: -20°C.