Learn More
Aviva Systems Biology Corp CSHL1 Antibody - C-terminal region (ARP41935_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41935_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSHL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CSHL1. Alternate names: CS-5, CSHP1, CSL, MGC149868, hCS-LApplicationWB. Species reactivity: Cow, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58056