Learn More
Cloud Clone Corp PRIMARY CELLS DOG SPRAD 5X10 5
Catalog No. NC3953283
Encompass_Preferred
Primary Canine Subcutaneous Preadipocytes (SPrAD) are normal primary cells derived from canine subcutaneous tissue. These cells are supplied in a 5X10^5 quantity, available in either fresh or cryopreserved formats for research applications.
  • Adherent growth characteristics
  • Cultured in DMEM/F12 supplemented with 5% FBS, 1% mesenchymal stem cell growth supplement, and 1% penicillin-streptomycin
  • Validated negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Provided in frozen stock containing 90% FBS and 10% DMSO or as a T25 flask culture
  • Certified under ISO 9001 and ISO 13485 quality management systems
Catalog No. NC3953283 Supplier Cloud Clone Corp Supplier No. CSI032CA01
Primary Canine Subcutaneous Preadipocytes (SPrAD) are normal primary cells derived from canine subcutaneous tissue. These cells are supplied in a 5X10^5 quantity, available in either fresh or cryopreserved formats for research applications.
  • Adherent growth characteristics
  • Cultured in DMEM/F12 supplemented with 5% FBS, 1% mesenchymal stem cell growth supplement, and 1% penicillin-streptomycin
  • Validated negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Provided in frozen stock containing 90% FBS and 10% DMSO or as a T25 flask culture
  • Certified under ISO 9001 and ISO 13485 quality management systems