Learn More
Cloud Clone Corp PRIMARY CELLS CANINE PF 5X10 5
Catalog No. NC3953285
Encompass_Preferred
Primary Canine Pulmonary Fibroblasts (PF) are derived from healthy lung tissue of Canis familiaris. These adherent primary cells are isolated and prepared to support various research applications. The cells are provided at a concentration of 5x10^5 cells/mL, maintaining an activity level of approximately 85%.
  • Adherent growth properties
  • Derived from normal canine lung tissue
  • Biosafety tested and confirmed negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Cultured in medium supplemented with 10% FBS, pulmonary fibroblasts supplement, and penicillin-streptomycin
  • Cryopreserved in a solution containing 90% FBS and 10% DMSO
  • For research use only; not for human or animal clinical diagnostic use
Catalog No. NC3953285 Supplier Cloud Clone Corp Supplier No. CSI034CA01
Primary Canine Pulmonary Fibroblasts (PF) are derived from healthy lung tissue of Canis familiaris. These adherent primary cells are isolated and prepared to support various research applications. The cells are provided at a concentration of 5x10^5 cells/mL, maintaining an activity level of approximately 85%.
  • Adherent growth properties
  • Derived from normal canine lung tissue
  • Biosafety tested and confirmed negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Cultured in medium supplemented with 10% FBS, pulmonary fibroblasts supplement, and penicillin-streptomycin
  • Cryopreserved in a solution containing 90% FBS and 10% DMSO
  • For research use only; not for human or animal clinical diagnostic use