Learn More
Cloud Clone Corp CSI047Ra01 Cloud-Clone Brand Primary Rat Meningeal Cells (MC) Size 5X10^5 Ship with dry ice
Catalog No. NC3953291
Encompass_Preferred
Species: Rat. Tissue Source: Cerebral pia mater. Growth Properties: Adherent. Cell Tpye: Meningeal Cells. Research Field: Nervous cell series;Meningeal cells;Cerebral and nervous systems;
Catalog No. NC3953291 Supplier Cloud Clone Corp Supplier No. CSI047RA01
Species: Rat. Tissue Source: Cerebral pia mater. Growth Properties: Adherent. Cell Tpye: Meningeal Cells. Research Field: Nervous cell series;Meningeal cells;Cerebral and nervous systems;