Learn More
Cloud Clone Corp CSI057Fe01 Cloud-Clone Brand Primary Feline Skeletal Muscle Cells (SkMC) Size 5X10^5 Ship with dry ice
Catalog No. NC3953292
Encompass_Preferred
Species: Cat. Tissue Source: Limb muscles tissue. Growth Properties: Adherent. Cell Tpye: Skeletal Muscle Cells. Research Field: Skeletal cell series;Skeletal muscle cellsSkeletal, articular and cutaneous systems
Catalog No. NC3953292 Supplier Cloud Clone Corp Supplier No. CSI057FE01
Species: Cat. Tissue Source: Limb muscles tissue. Growth Properties: Adherent. Cell Tpye: Skeletal Muscle Cells. Research Field: Skeletal cell series;Skeletal muscle cellsSkeletal, articular and cutaneous systems