Learn More
Cloud Clone Corp CSI065Rb01 Cloud-Clone Brand Primary Rabbit Bladder Smooth Muscle Cells (BSMC) Size 5X10^5 Ship with dry ice
Catalog No. NC3953295
Encompass_Preferred
Species: Rabbit. Tissue Source: Bladder tissue. Growth Properties: Adherent. Cell Tpye: Smooth Muscle Cells. Research Field: Urethral cell series;Smooth muscle cells;Urinary system;
Catalog No. NC3953295 Supplier Cloud Clone Corp Supplier No. CSI065RB01
Species: Rabbit. Tissue Source: Bladder tissue. Growth Properties: Adherent. Cell Tpye: Smooth Muscle Cells. Research Field: Urethral cell series;Smooth muscle cells;Urinary system;