Learn More
Cloud Clone Corp CANINE TSMC 5X10 5
Catalog No. NC3953297
Encompass_Preferred
Primary Canine Tracheal Smooth Muscle Cells (TSMC) are isolated from canine tracheal tissue and provided as an adherent cell culture. These cells are tested and confirmed negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.
  • Supplied as a frozen 1 mL aliquot (90% FBS, 10% DMSO) or in a T25 flask
  • Requires culture in DMEM supplemented with 5% FBS, 1% smooth muscle cell growth supplement, and 1% penicillin-streptomycin
  • Maintained at 37℃ in a 95% air and 5% carbon dioxide environment
  • Validated for research use only, not for human or animal diagnostic or clinical applications
  • Cells demonstrate 85% activity and exhibit adherent growth properties
  • Storage requires immediate transfer to liquid nitrogen upon receipt
Catalog No. NC3953297 Supplier Cloud Clone Corp Supplier No. CSI068CA01
Primary Canine Tracheal Smooth Muscle Cells (TSMC) are isolated from canine tracheal tissue and provided as an adherent cell culture. These cells are tested and confirmed negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi.
  • Supplied as a frozen 1 mL aliquot (90% FBS, 10% DMSO) or in a T25 flask
  • Requires culture in DMEM supplemented with 5% FBS, 1% smooth muscle cell growth supplement, and 1% penicillin-streptomycin
  • Maintained at 37℃ in a 95% air and 5% carbon dioxide environment
  • Validated for research use only, not for human or animal diagnostic or clinical applications
  • Cells demonstrate 85% activity and exhibit adherent growth properties
  • Storage requires immediate transfer to liquid nitrogen upon receipt