Learn More
Cloud Clone Corp CANINE BMMSCS 5X10 5
Catalog No. NC3953298
Encompass_Preferred
Primary Canine Bone Marrow-derived Mesenchymal Stem Cells (BMMSCs) are isolated from beagle dog bone marrow and provided as an adherent primary cell population. These cells are designed for laboratory research applications and are screened to be negative for pathogens, ensuring high-quality performance in controlled experimental environments.
  • Derived from canine bone marrow
  • Adherent growth characteristics
  • Pathogen-free including negative results for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • High viability of >85%
  • Minimum of 5x10^5 cells per vial
  • Supplied in frozen stock solution containing 90% FBS and 10% DMSO or as a T25 flask culture
  • Requires DMEM/F12-based medium supplemented with growth factors and antibiotics
Catalog No. NC3953298 Supplier Cloud Clone Corp Supplier No. CSI083CA01
Primary Canine Bone Marrow-derived Mesenchymal Stem Cells (BMMSCs) are isolated from beagle dog bone marrow and provided as an adherent primary cell population. These cells are designed for laboratory research applications and are screened to be negative for pathogens, ensuring high-quality performance in controlled experimental environments.
  • Derived from canine bone marrow
  • Adherent growth characteristics
  • Pathogen-free including negative results for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • High viability of >85%
  • Minimum of 5x10^5 cells per vial
  • Supplied in frozen stock solution containing 90% FBS and 10% DMSO or as a T25 flask culture
  • Requires DMEM/F12-based medium supplemented with growth factors and antibiotics