Learn More
Cloud Clone Corp CSI083Rb01 Cloud-Clone Brand Primary Rabbit Bone Marrow-derived Mesenchymal Stem Cells (BMMSCs) Size 5X10^5 Ship with dry ice
Catalog No. NC3953300
Encompass_Preferred
Species: Rabbit. Tissue Source: Bone marrow. Growth Properties: Adherent. Cell Tpye: Mesenchymal Stem Cells. Research Field: Mesenchymal stem cell series;Mesenchymal stem cells;Skeletal, articular and cutaneous systems;
Catalog No. NC3953300 Supplier Cloud Clone Corp Supplier No. CSI083RB01
Species: Rabbit. Tissue Source: Bone marrow. Growth Properties: Adherent. Cell Tpye: Mesenchymal Stem Cells. Research Field: Mesenchymal stem cell series;Mesenchymal stem cells;Skeletal, articular and cutaneous systems;