Learn More
Cloud Clone Corp CSI102Ra01 Cloud-Clone Brand Primary Rat Osteoblasts (OB) Size 5X10^5 Ship with dry ice
Supplier:  Cloud Clone Corp CSI102RA01
Encompass_Preferred
Species: Rat. Tissue Source: Neonatal rat calvarium bone. Growth Properties: Adherent. Cell Tpye: Osteoblast. Research Field: Skeletal cell series;Osteoblasts;Skeletal, articular and cutaneous systems;
Catalog No. NC3953305