Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloud Clone Corp PRIMARY CELLS DOG GSMC 5X10 5

Catalog No. NC3953306
Encompass_Preferred

Primary Canine Gastric Smooth Muscle Cells (GSMC) are isolated from normal canine gastric tissue for research applications. These adherent cells are suitable for gastrointestinal and digestive system studies and are provided in a quantity of 5x10^5 cells per unit.

  • Derived from Canis familiaris species
  • Growth properties: adherent
  • Culture requirements: DMEM supplemented with 5% FBS, 1% smooth muscle cell growth supplement, and 1% penicillin-streptomycin solution
  • Incubation conditions: 37°C in a 5% carbon dioxide atmosphere
  • Biosafety verified: negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Format: available as frozen 1 mL or T25 flask

Catalog No. NC3953306 Supplier Cloud Clone Corp Supplier No. CSI109CA01
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Primary Canine Gastric Smooth Muscle Cells (GSMC) are isolated from normal canine gastric tissue for research applications. These adherent cells are suitable for gastrointestinal and digestive system studies and are provided in a quantity of 5x10^5 cells per unit.

  • Derived from Canis familiaris species
  • Growth properties: adherent
  • Culture requirements: DMEM supplemented with 5% FBS, 1% smooth muscle cell growth supplement, and 1% penicillin-streptomycin solution
  • Incubation conditions: 37°C in a 5% carbon dioxide atmosphere
  • Biosafety verified: negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Format: available as frozen 1 mL or T25 flask

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.