Learn More
Cloud Clone Corp CSI113Ra01 Cloud-Clone Brand Primary Rat Nucleus Pulposus Cells (NPC) Size 5X10^5 Ship with dry ice
Catalog No. NC3953309
Encompass_Preferred
Species: Rat. Tissue Source: Intervertebral disc. Growth Properties: Adherent. Cell Tpye: Nucleus Pulposus Cells. Research Field: Skeletal cell series;Nucleus pulposus cells;Skeletal, articular and cutaneous systems;
Catalog No. NC3953309 Supplier Cloud Clone Corp Supplier No. CSI113RA01
Species: Rat. Tissue Source: Intervertebral disc. Growth Properties: Adherent. Cell Tpye: Nucleus Pulposus Cells. Research Field: Skeletal cell series;Nucleus pulposus cells;Skeletal, articular and cutaneous systems;