Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cloud Clone Corp PRIMARY CELLS RAT TGN 5X10 5

Catalog No. NC3953323
Encompass_Preferred

Primary Rat Trigeminal Ganglion Neuron Cells are isolated from Rattus norvegicus (SD rats) and provided as a frozen stock in 90% FBS and 10% DMSO. These primary neuron cells are suitable for neurological research applications involving trigeminal ganglion studies and are validated for purity via immunofluorescence identification of β-TubulinIII.

  • Adherent cell type
  • Species: Rattus norvegicus
  • Viability: 85% by trypan blue exclusion
  • Recommended culture conditions: 37 °C in 95% air and 5% CO2
  • Special culture medium: Neurobasal-A medium, B-27 supplement (50X), and 1% penicillin-streptomycin solution
  • Tested negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Pack size: 5x10^5 cells per vial
  • Non-proliferating; not recommended for expansion or long-term culture

Catalog No. NC3953323 Supplier Cloud Clone Corp Supplier No. CSI238RA01
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Primary Rat Trigeminal Ganglion Neuron Cells are isolated from Rattus norvegicus (SD rats) and provided as a frozen stock in 90% FBS and 10% DMSO. These primary neuron cells are suitable for neurological research applications involving trigeminal ganglion studies and are validated for purity via immunofluorescence identification of β-TubulinIII.

  • Adherent cell type
  • Species: Rattus norvegicus
  • Viability: 85% by trypan blue exclusion
  • Recommended culture conditions: 37 °C in 95% air and 5% CO2
  • Special culture medium: Neurobasal-A medium, B-27 supplement (50X), and 1% penicillin-streptomycin solution
  • Tested negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi
  • Pack size: 5x10^5 cells per vial
  • Non-proliferating; not recommended for expansion or long-term culture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.