Learn More
Carnabio Usa Inc CSK/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 081115
Encompass
CSK, Full-length, Wild type, Amino Acid 1-450, NP_004374.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0388