Learn More
Eurofins Discoverx CSK, active, 10 µg. (14-458)
Supplier:  Eurofins Discoverx 14458
Encompass
Recombinant, human full length CSK, containing an N-terminal GST-tag incorporating a PreScission Protease cleavage site. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8819