Learn More
Aviva Systems Biology Corp CSK Antibody - middle region (ARP52042_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52042_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSK. Alternate names: MGC117393ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-63043