Learn More
Enzo Life Sciences Csk (human), (recombinant) (GST-tag) (5 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE3800005
Encompass_Preferred
Formulation: Liquid. In 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol. Source: Produced in insect cells. Full length active enzyme. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2005