Learn More
Medchemexpress LLC Csk-in-1 | 2380027-49-4 | 98.6% | C34H32ClFN8O4 | 10MG
SDP
Catalog No. 5000260076
Encompass_Preferred
CSK-IN-1 is a potent, orally active inhibitor of C-terminal Src kinase (CSK) provided as a powder for research use. It demonstrates low-nanomolar biochemical potency and has been reported to enhance T cell activation and proliferation, and to reduce inhibitory LCK phosphorylation in vivo. Recommended storage conditions are provided for powder and solution forms.
Catalog No. 50-002-60076 Supplier Medchemexpress LLC Supplier No. HY13025410MG
CSK-IN-1 is a potent, orally active inhibitor of C-terminal Src kinase (CSK) provided as a powder for research use. It demonstrates low-nanomolar biochemical potency and has been reported to enhance T cell activation and proliferation, and to reduce inhibitory LCK phosphorylation in vivo. Recommended storage conditions are provided for powder and solution forms.