Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-Ade-His-Leu-Lys-Trp-Ura (Powder)

Catalog No. MP114560122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114560122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114560122 Supplier MP Biomedicals™ Supplier No. 114560122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Time tested formulation. MP Biomedicals CSM-Ade-His-Leu-Lys-Trp-Ura (Powder) is a 4+ dropout of Complete Supplement Mixture of amino acids for S. cerevisiae growth media. Suitable for use with yeast cell culture, gene expression and transcription regulati

To use, add 0.54g of CSM-Ade-His-Leu-Lys-Trp-Ura to each liter of DOB or DOBA. Then, autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Yeast cell culture, Gene Expression, Transcription Regulatiotn Research
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.