Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-Arg (Powder)

Catalog No. MP114510122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114510122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114510122 Supplier MP Biomedicals™ Supplier No. 114510122
Add to Cart
Add to Cart

Custom formulate your own selective Synthetic Defined yeast media. MP Biomedicals CSM-Arg (Powder) is a single dropout of Complete Supplement Mixture of amino acids for S. cerevisiae growth media. For use with DOB or YNB with the appropriate nitrogen and

Single Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments. Add 0.74g of CSM-Arg to each liter of purified water and DOB or DOBA. Then, autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Single Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments.
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.