Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-His-Leu-Met-Trp-Ura (Powder)

Catalog No. MP114550422 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114550422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114550422 Supplier MP Biomedicals™ Supplier No. 114550422
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Custom formulate your own selective Synthetic Defined yeast media. MP Biomedicals CSM-His-Leu-Met-Trp-Ura (Powder) is a 5 dropout of Complete Supplement Mixture of amino acids for S. cerevisiae growth media. For use with DOB or YNB with the appropriate ni

5 Dropout Complete Supplement Mixture is used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments. Reagents are tested and chosen for their ability to promote optimum growth of yeast strains when combined in the DOB or DOBA with this formula.

Chosen reagents are combined in the proper proportions according to the formulation listed in the certificate of analysis and milled to a powder which thoroughly blends the reagents for uniform distribution.

  • A sample of the milled and blended formulation was used to prepare liquid plate medium
  • 43.7g DOBA powder and 0.56g of CSM-His-Leu-Met-Trp-Ura were added to 1L of purified water
  • The combinations were mixed by hand for about 1 minute to dissolve the dextrose
  • The mixture was then autoclaved at 121°C for 25 minutes
  • After cooling to 50°C, plates were poured
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) 5 Dropout Complete Supplement Mixture used to select for yeast containing plasmids with specific nutritional markers and to select for gene function in transformation experiments.
Quantity 100 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.