Learn More
Sunrise Science Products CSM-Trp Powder, 10 grams
Catalog No. NC1408939
Encompass
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media. Mix with stirring for 10-15 minutes, and filter sterilize or autoclave at 121°C for 15 minutes. Recipe in mg/L: Adenine hemisulfate (10 mg/L), L-Arginine (50 mg/L), L-Aspartic acid (80 mg/L), L-Histidine hydrochloride monohydrate (20 mg/L), L-Isoleucine (50 mg/L), L-Leucine (100 mg/L), L-Lysine hydrochloride (50 mg/L), L-Methionine (20 mg/L), L-Phenylalanine (50 mg/L), L-Threonine (100 mg/L), L-Tyrosine (50 mg/L), L-Valine (140 mg/L), Uracil (20 mg/L).
Catalog No. NC1408939 Supplier Sunrise Science Products Supplier No. 1007010
Commonly added to YNB, nitrogen and glucose, at suggested g/L, for a complete yeast media. Mix with stirring for 10-15 minutes, and filter sterilize or autoclave at 121°C for 15 minutes. Recipe in mg/L: Adenine hemisulfate (10 mg/L), L-Arginine (50 mg/L), L-Aspartic acid (80 mg/L), L-Histidine hydrochloride monohydrate (20 mg/L), L-Isoleucine (50 mg/L), L-Leucine (100 mg/L), L-Lysine hydrochloride (50 mg/L), L-Methionine (20 mg/L), L-Phenylalanine (50 mg/L), L-Threonine (100 mg/L), L-Tyrosine (50 mg/L), L-Valine (140 mg/L), Uracil (20 mg/L).