Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-Trp with 20mg/L ADE (Powder)

Catalog No. MP114512522 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114512522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114512522 Supplier MP Biomedicals™ Supplier No. 114512522
Add to Cart
Add to Cart

Time tested formulation. MP Biomedicals CSM-Trp with 20 mg/L Ade (Powder) is a single dropout of Complete Supplement Mixture of amino acids for S. cerevisiae growth media. Suitable for use with yeast cell culture, gene expression and transcription regulat

To use, add 0.76g of CSM-Trp+Ade to each liter of purified water and DOB or DOBA. Then, autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
For Use With (Application) Yeast cell culture, Gene Expression, Transcription Regulatiotn Research
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.