Learn More
Aviva Systems Biology Corp CSN3 Antibody - N-terminal region (ARP52064_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52064_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSN3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CSN3. Alternate names: CSN10, CSNK, KCAApplicationWB. Species reactivity: Human
Catalog No. 50-285-629