Learn More
Aviva Systems Biology Corp CSNK1E Antibody - N-terminal region (ARP51987_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51987_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSNK1E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CSNK1E. Alternate names: HCKIE, MGC10398, CKIepsilonApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-60662