Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CSNK1E (CK1 ε), His Tag Recombinant Protein

Catalog No. PV3500
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3500 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3500 Supplier Invitrogen™ Supplier No. PV3500
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

CSNK1E (CK1 epsilon) is a serine/threonine kinase implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis. CSNK1E phosphorylate key regulatory proteins involved in the control of cell differentiation, proliferation, chromosome segregation, circadian rhythms, and metabolic pathways. CK1 family members share a highly conserved kinase domain but differ in their variable N-and C-terminal domains. Mammals have seven family members: alpha, beta, gamma1, gamma2, gamma3, delta and epsilon. CKIe positively regulates Akt possibly by inhibiting PP2A. CKIe play an important role in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49674
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 4mM DTT, 50% glycerol, 0.04% Triton X-100, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 1454
Molecular Weight (g/mol) 52.3 kDa
Name Human CSNK1E (CK1 ε), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI426939; AI551861; AW457082; casein kinase 1; casein kinase 1 epsilon; casein kinase 1, epsilon; casein kinase I isoform epsilon; CK1e; CK1epsilon; CKI, epsilon; CKIe; CKIepsilon; CKI-epsilon; Csnk1e; HCKIE; KC1 epsilon; KC1epsilon; MGC10398; OTTHUMP00000063258; OTTHUMP00000063263; RP1-5O6.1; tau
Common Name CK1 epsilon
Gene Symbol CSNK1E
Kinase Family CK1 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.04% Triton¤ X-100, 4 mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CK1 Group
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™, Omnia™ Bulk Substrate
Target Entry CSNK1E
Target Gene casein kinase 1, epsilon
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.