Learn More
Aviva Systems Biology Corp CSNK1G1 Antibody - middle region (ARP37531_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37531_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CSNK1G1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse CSNK1G1. Alternate names: 9130020E21RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56380