Learn More
Aviva Systems Biology Corp Csnk1g1 Antibody - middle region (ARP37530_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37530_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Csnk1g1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Csnk1g1. Alternate names: 9130020E21RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-56379