Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CSNK1G3 (CK1 gamma 3), GST Tag Recombinant Protein

Catalog No. PV3838
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3838 10 μg
PR7474A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PV3838 Supplier Invitrogen™ Supplier No. PV3838
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6M4
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 2mM DTT, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 1456
Molecular Weight (g/mol) 78.9 kDa
Name Human CSNK1G3 (CK1 gamma 3), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L
Common Name CK1 gamma-3
Gene Symbol CSNK1G3
Kinase Family CK1 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CK1 Group
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™, Omnia™ Bulk Substrate
Target Entry CSNK1G3
Target Gene casein kinase 1, gamma 3
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Purity or Quality Grade 80% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.