Learn More
Aviva Systems Biology Corp CSNK2A1 Antibody - middle region (ARP51990_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51990_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSNK2A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSNK2A1. Alternate names: CK2A1, CKII, CKII alphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60663