Learn More
Aviva Systems Biology Corp CSNK2A2 Antibody - C-terminal region (ARP53596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSNK2A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CSNK2A2. Alternate names: CK2A2, CSNK2A1, FLJ43934ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61063