Learn More
Enzo Life Sciences CSP polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAPSV003E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of rat CSP. The sequence is completely conserved in bovine. UniProt: P60905. Source: Purified from rabbit serum. Species Reactivity: Mouse, Rat, Xenopus, Bovine. Applications: IHC, IP, WB. Recommended dilutions: Immunohistochemistry, Immunoprecipitation (0.5µg/ml). Western Blot (1:2,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-875