Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CSP1 | 172889-49-5 | 98.3% | 2242.69 | 10 MG
SDP

Catalog No. 5000424280
Encompass_Preferred

CSP1 is a potent and selective ComD1 receptor agonist, with an IC50 of 10.3 nM. It is a major variant of competence-stimulating peptide (CSP) and regulates genetic transformation of S. pneumonia by modulating quorum sensing (QS). CSP1 can also act as an antibacterial agent.

  • Solid, white to off-white appearance
  • Sequence: Glu-Met-Arg-Leu-Ser-Lys-Phe-Phe-Arg-Asp-Phe-Ile-Leu-Gln-Arg-Lys-Lys
  • Soluble in H2O: 100 mg/mL (44.59 mM); requires ultrasonic
  • Soluble in PBS: 50 mg/mL (22.29 mM); clear solution; requires ultrasonic
  • Target: Bacterial
  • Pathway: Anti-infection

Catalog No. 50-004-24280 Supplier Medchemexpress LLC Supplier No. HYP245410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CSP1 is a potent and selective ComD1 receptor agonist, with an IC50 of 10.3 nM. It is a major variant of competence-stimulating peptide (CSP) and regulates genetic transformation of S. pneumonia by modulating quorum sensing (QS). CSP1 can also act as an antibacterial agent.

  • Solid, white to off-white appearance
  • Sequence: Glu-Met-Arg-Leu-Ser-Lys-Phe-Phe-Arg-Asp-Phe-Ile-Leu-Gln-Arg-Lys-Lys
  • Soluble in H2O: 100 mg/mL (44.59 mM); requires ultrasonic
  • Soluble in PBS: 50 mg/mL (22.29 mM); clear solution; requires ultrasonic
  • Target: Bacterial
  • Pathway: Anti-infection

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.