Learn More
Medchemexpress LLC CSP1 | 172889-49-5 | 98.3% | 2242.69 | 5 MG
SDP
Catalog No. 5000424165
Encompass_Preferred
CSP1 is a potent and selective ComD1 receptor agonist with an IC50 of 10.3 nM. It is a major variant of competence-stimulating peptide (CSP) that regulates the genetic transformation of *S. pneumonia* by modulating quorum sensing (QS). CSP1 can also act as an antibacterial agent.
  • Potent and selective ComD1 receptor agonist
  • Regulates genetic transformation of *S. pneumonia*
  • Modulates quorum sensing (QS)
  • Acts as an antibacterial agent
Catalog No. 50-004-24165 Supplier Medchemexpress LLC Supplier No. HYP24545MG
CSP1 is a potent and selective ComD1 receptor agonist with an IC50 of 10.3 nM. It is a major variant of competence-stimulating peptide (CSP) that regulates the genetic transformation of *S. pneumonia* by modulating quorum sensing (QS). CSP1 can also act as an antibacterial agent.
  • Potent and selective ComD1 receptor agonist
  • Regulates genetic transformation of *S. pneumonia*
  • Modulates quorum sensing (QS)
  • Acts as an antibacterial agent