Learn More
Sigma Aldrich Fine Chemicals Biosciences CSPD ready to use Disodium
Catalog No. 502905248
Encompass
CSPD is a chemiluminescent substrate for alkaline phosphatase that enables extremely fast and sensitive detection of biomolecules by producing visible light. Light emission is recorded on X-ray film or on luminescence imager systems.
Catalog No. 50-290-5248 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11755633001
CSPD is a chemiluminescent substrate for alkaline phosphatase that enables extremely fast and sensitive detection of biomolecules by producing visible light. Light emission is recorded on X-ray film or on luminescence imager systems.