Learn More
Aviva Systems Biology Corp CSPG5 Antibody - N-terminal region (ARP46737_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46737_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSPG5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CSPG5. Alternate names: MGC44034, NGCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59388