Learn More
Med Vet International Dynarex Csr Wraps, 54" X 72", 100/Case
Catalog No. 502828956
Encompass_Preferred
Guarantee sterility with this superior, tear-resistant wrap. Superior, non-woven, wrapping fabric for steam and ethylene oxide (EO) gas sterilization Folds and drapes like muslin - no spring-back Repels water, blood, and saline Strong, tear-resistant material Effective barrier against air and water-borne bacteria
Catalog No. 50-282-8956 Supplier Med Vet International Supplier No. 4472
Guarantee sterility with this superior, tear-resistant wrap. Superior, non-woven, wrapping fabric for steam and ethylene oxide (EO) gas sterilization Folds and drapes like muslin - no spring-back Repels water, blood, and saline Strong, tear-resistant material Effective barrier against air and water-borne bacteria