Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CSRM617 hydrochloride | 1353749-74-2 | 100.0% | 100 MG
SDP

Catalog No. 5000232039
Encompass_Preferred

CSRM617 hydrochloride is the hydrochloride salt of a selective small-molecule inhibitor of the transcription factor ONECUT2, supplied as a white to off-white solid for research use.

  • Selective inhibitor of ONECUT2 transcription factor.
  • Hydrochloride salt, white to off-white solid.
  • Purity 99.97%.
  • Molecular weight 291.69 g/mol.
  • Soluble in DMSO (100 mg/mL; ultrasonic assistance recommended).
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human use.

Catalog No. 50-002-32039 Supplier Medchemexpress LLC Supplier No. HY122611A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CSRM617 hydrochloride is the hydrochloride salt of a selective small-molecule inhibitor of the transcription factor ONECUT2, supplied as a white to off-white solid for research use.

  • Selective inhibitor of ONECUT2 transcription factor.
  • Hydrochloride salt, white to off-white solid.
  • Purity 99.97%.
  • Molecular weight 291.69 g/mol.
  • Soluble in DMSO (100 mg/mL; ultrasonic assistance recommended).
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.