Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Csrm617 hydrochloride | 1353749-74-2 | 100.0% | 25 MG
SDP

Catalog No. 5000230950
Encompass_Preferred

CSRM617 hydrochloride is a selective small-molecule inhibitor of the transcription factor ONECUT2 (OC2). It binds the OC2-HOX domain (Kd 7.43 μM in SPR assays), induces apoptosis in preclinical models, and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Selective ONECUT2 inhibitor with documented apoptosis induction.
  • High purity: 99.97%.
  • Available in multiple solid pack sizes including 25 MG.
  • Soluble in DMSO: 100 mg/mL; several in vivo formulation protocols provided.
  • Storage: solid at -20°C; solutions at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-30950 Supplier Medchemexpress LLC Supplier No. HY122611A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CSRM617 hydrochloride is a selective small-molecule inhibitor of the transcription factor ONECUT2 (OC2). It binds the OC2-HOX domain (Kd 7.43 μM in SPR assays), induces apoptosis in preclinical models, and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Selective ONECUT2 inhibitor with documented apoptosis induction.
  • High purity: 99.97%.
  • Available in multiple solid pack sizes including 25 MG.
  • Soluble in DMSO: 100 mg/mL; several in vivo formulation protocols provided.
  • Storage: solid at -20°C; solutions at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.