Learn More
Aviva Systems Biology Corp CSRP1 Antibody - middle region (ARP34391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSRP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSRP1. Alternate names: CRP, CRP1, CSRP, CYRP, D1S181E, DKFZp686M148ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55374