Learn More
Aviva Systems Biology Corp CSRP3 Antibody - middle region (ARP34181_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34181_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CSRP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSRP3. Alternate names: CLP, MLP, CRP3, LMO4, CMD1M, CMH12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55286