Learn More
Electron Microscopy Sciences IP Biopsy VI IP Biopsy VI Cassettes, Taped White
SDP
Catalog No. 501926939
Encompass_Preferred
IP Biopsy VI cassettes are two-piece biopsy cassettes with 0.67 mm pores, a 45 degree writing surface and a secure latch closure. Taped cassettes have been designed for the Leica IPC Cassette Printer and offer <1% printing error rate. Taped: CE approved weight for IPC Printers-4.5 Kg, dimensions inches (L x W x H)-17 x 11 x 7.
Color: Taped White
This item is not returnable. View return policy
Catalog No. 50-192-6939 Supplier Electron Microscopy Sciences Supplier No. 70076111
IP Biopsy VI cassettes are two-piece biopsy cassettes with 0.67 mm pores, a 45 degree writing surface and a secure latch closure. Taped cassettes have been designed for the Leica IPC Cassette Printer and offer <1% printing error rate. Taped: CE approved weight for IPC Printers-4.5 Kg, dimensions inches (L x W x H)-17 x 11 x 7.
Color: Taped White