Learn More
Electron Microscopy Sciences ActivFlo Mini Biopsy Cassettes, Tan
SDP
Catalog No. 501926967
Encompass_Preferred
Activflo Mini Cassettes are one third the size of standard cassettes, and feature sloped walls, creating a basket for small tissue management. The smaller size allows double the number of blocks to be stored, reducing specimen storage space. Excellent solution for microwave and rapid tissue processors.
Color: Tan
This item is not returnable. View return policy
Catalog No. 50-192-6967 Supplier Electron Microscopy Sciences Supplier No. 70076137
Activflo Mini Cassettes are one third the size of standard cassettes, and feature sloped walls, creating a basket for small tissue management. The smaller size allows double the number of blocks to be stored, reducing specimen storage space. Excellent solution for microwave and rapid tissue processors.
Color: Tan