Learn More
Aviva Systems Biology Corp Cst6 Antibody - middle region (ARP53533_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53533_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cst6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human Cst6. Alternate names: 1110017E11Rik, N28197, ichqApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-847