Learn More
Aviva Systems Biology Corp CST8 Antibody - middle region (ARP52079_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52079_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CST8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CST8. Alternate names: CRESApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53981